missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester GHS Symbol Signs

Help ensure safety at work and maximize productivity with these GHS warning labels. Brady™ Laminated Polyester GHS Symbol Signs are available in a variety of sizes, with and without text. Standardized images reduce the risk of misunderstandings.

Brand:  Brady™ 814037

118.75 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15664237

  • £127.00 / Pack of 250
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Self-adhesive diamond shape labels
  • Pre-printed with GHS hazard symbols
  • Laminated polyester, gloss finish black and red on white
Specifications

Specifications

ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Polyester
Individual Markers on a Roll
Laminated Polyester
50 mm
50 mm
Diamond
GHS Symbol Sign
Gloss
250/Pk.
Permanent Acrylic
Pipe markers and safety signs
0.024 mm
-40°C to 110°C
Hazardous to Aquatic Environment
Black on White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers