missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester: ISO Safety Sign - No stepping on surface

Sign compliant to ISO 7010.

Brand:  Brady™ 824232

 View more versions of this product

Product Code. 15674637

  • £4.50 / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Laminated polyester
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
Polyester
Permanent Acrylic
Indoor and Outdoor
-40°C to 110°C
Global
Gloss
1 Ea.
200 mm
0.099 mm
Round
Red
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers