Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Use this walkway
Sign compliant to ISO 7010.
Brand:  Brady™ 821400
 View more versions of this product
Product Code. 15654557
  • £3.50 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
Gloss
1 Ea.
100 mm
0.099 mm
Round
White on Blue
PIC M024-Dia 100-PE-CRD/1
Polyester
Permanent Acrylic
Indoor and Outdoor
-40°C to 110°C
Global