missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester: ISO Safety Sign - Wear safety footwear

Sign compliant to ISO 7010.

Brand:  Brady™ 819016

3.14 GBP valid until 2024-09-30
Use promo code "21675" to get your promotional price.


 View more versions of this product

Product Code. 15614477

  • £3.30 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Laminated polyester
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
Polyester
Permanent Acrylic
Indoor and Outdoor
-40°C to 110°C
Global
Gloss
1 Ea.
100 mm
0.099 mm
Round
White on Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documentos

Documentos

Product Certifications
Ofertas especiales

Ofertas especiales