missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester Stickers

Brand:  Brady™ 227588

 View more versions of this product

Product Code. 11770254

  • £4.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

ISO 7010
Polyester
Permanent Acrylic
Pipemarkers and safety signs
0.024 mm
-40°C to 110°C
Flammable Gases 2
Black on Red
Gloss
1 Ea.
Laminated Polyester
200 mm
200 mm
Rhomboid
Dangerous Goods Transport Label
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers