missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Laminated Polyester Warning Safety Signs
£103495.00
Specifications
Certifications/Compliance | ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance |
---|---|
Material | Polyester |
Quantity | 1 Ea. |
Description | Laminated Polyester |
For Use With (Application) | Pipe markers and safety signs |
Product Code | Brand | Displayed Text | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Width (Metric) | Price | Quantity & Availability | ||||
10151984
|
Brady™
250273 |
Warning about skid risk | 100 mm |
£103495.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041344
|
Brady™
250225 |
Danger Zone Warning Pictogram - ! | 100 mm |
£103495.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10264894
|
Brady™
250226 |
Danger Zone Warning Pictogram - ! | 200 mm |
£103495.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10441074
|
Brady™
250219 |
Warning of industrial trucks | 100 mm |
£103495.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10061394
|
Brady™
250276 |
Warning of tripping hazard | 100 mm |
£103495.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance | |
1 Ea. | |
Pipe markers and safety signs | |
Triangular | |
Black on Yellow |
Polyester | |
Laminated Polyester | |
0.024 mm | |
Warning Safety Sign |