Learn More
Gibco™ Laminin Mouse Protein, Natural
Laminin Mouse Protein, Natural
Brand: Gibco™ 23017015
172.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Laminin is the major glycoprotein component of basement membranes, and has a molecular weight of 850,000 daltons. This preparation of mouse Laminin was isolated from the Engelbreth-Holm-Swarm (EHS) sarcoma and is supplied at a concentration of 0.5-2.0 mg⁄ml.Key functions include:
Supplied: 1 mg in 50 mM Tris-HCL (pH 7.4), 0.15 M NaCl
Performance qualification (cell attachment): Half-maximal cell attachment of MG63 cells at 37°C: ≤ 20 ug⁄ml

Specifications
Cell Culture | |
Laminin | |
Store in ultra-cold freezer (-68 to -85°C) and protect from light. | |
Liquid | |
Extracellular Matrix & Cell Adhesion, Stem Cell Research |
1 mg | |
Mouse | |
Dry Ice | |
Laminin |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3041333 | Certificate of Analysis | 31/01/2025 | 23017015 |
3042837 | Certificate of Analysis | 31/01/2025 | 23017015 |
3018303 | Certificate of Analysis | 31/01/2025 | 23017015 |
3082246 | Certificate of Analysis | 31/01/2025 | 23017015 |
3047783 | Certificate of Analysis | 31/01/2025 | 23017015 |
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
38Hours
4Minutes
0Seconds
10New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.