missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 218.0°C |
Loss on Drying | 0.5% max. (105°C, 3 h) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in dmf |
Formula Weight | 256.09 |
Percent Purity | 98% |
Chemical Name or Material | Lamotrigine |
Safety and Handling

Product Identifier
- Lamotrigine, 98%
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- MIXTURE LIST-Contains : Lamotrigine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lamotrigine, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction