missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips
GREENER_CHOICE

Product Code. p-3256647 Shop All Cole-Parmer Products
Click to view available options
Parameter Specifications:
Chlorine dioxide
Peracetic acid
Quantity:
50 Strips/Vial
Range:
0 to 500 ppm
0-10-20-40-60-85-160 ppm
Unit Size:
100µL
Each
This item is not returnable. View return policy

Product Code. 13423717

Brand: LaMotte™ PA529558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For water quality testing

  • No reagents or field kits required for most strips
  • Chemically treated pads on plastic strips produce color change
  • Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
  • As little as 10-second development time

Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †

†Strips have been evaluated for use in these applications. Use in other applications is subject to potential interferences.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Water quality testing
Parameter Specifications Peracetic acid
Quantity 50 Strips/Vial
Range 0-10-20-40-60-85-160 ppm
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Type Single Factor Test Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.