missing translation for 'onlineSavingsMsg'
Learn More

3M™ Lane and Safety Marking Tape 471

Product Code. 17781679
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Unit Size:
12 rolls
This item is not returnable. View return policy

Product Code. 17781679

Brand: 3M™ 7100260125

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Strength, conformability, instant adhesion and long-term clean removal make this tape an ideal choice for all but the heaviest floor and safety marking applications. Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, and splicing, this tape comes individually wrapped and conveniently packaged.

  • Maintains color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, high holding strength on many surfaces
  • Available in many colors (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Clean removal from many surfaces to help reduce clean-up and labor costs
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 33 m x 50 mm
Packaging Quantity 12 Rolls
Thickness 0.14 mm
Adhesive Type Rubber-based Adhesive
Color Red
Material Vinyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.