missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum boride, 99.5% (REO), Thermo Scientific Chemicals Product Code.: 11393888

Lanthanum boride, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 11393888
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11393888

Brand: Thermo Scientific Alfa Aesar 040325.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum boride is employed in hot cathodes, which enact and includes electron microscopes, microwave tubes, electron lithography, electron beam welding, X-ray tubes, and free electron lasers. It is administered as a size or strain standard in X-ray powder diffraction to calibrate instrumental broadening of diffraction peaks. It exercises as a superconductor. It has a great potential as a thermionic emitter in renewable energy applications. It is widely used in teleran, electronic industry, instrument, medical equipment, metallurgy and environmental protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12008-21-8
Molecular Formula B6La-2
Molecular Weight (g/mol) 203.765
MDL Number MFCD00151350
InChI Key IBDHMAYMLSYGAK-UHFFFAOYSA-N
Synonym lanthanum boride, lanthanum hexaboride, lanthanum boride reo, lanthanum hexaboride lab6 grade a, lanthanum hexaboride lab6 grade b, lanthanum hexaboride lab6 grade c, lanthanum hexaboride, powder, 10 mum, lanthanum boride, powder,-325 mesh trace metals basis, pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide lanthanum, line position and line shape standard for powder diffraction, nist srm 660c, lanthanum hexaboride powder
PubChem CID 71308229
SMILES B12B3[B-]14B5[B-]23B45.[La]

Specifications

Melting Point 2210°C
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water,HCl.
Formula Weight 203.78
Percent Purity 99.5%
Odor Odorless
Density 2.61 g/mL
Assay (REO)
Chemical Name or Material Lanthanum boride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN27L042 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number040325.09,040325.18,040325.30
Lot NumberR02K028 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number040325.09,040325.18,040325.30
Lot NumberC09X015 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040325.09,040325.18,040325.30
Lot NumberA05S006 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040325.09,040325.18,040325.30
Lot NumberA11W010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040325.09,040325.18,040325.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum boride, 99.5% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.