missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride heptahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Lanthanum chloride heptahydrate, 99.99%, Cl3La.7H2O, CAS Number-10025-84-0, 500g, White, 371.38, Trace Metal Basis, GHS Signal Word: Danger

£93.47 - £392.96

Chemical Identifiers

CAS 10025-84-0
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10762621
View Documents Promotion Details
Thermo Scientific Acros
193271000
100 g Glass bottle
£93.47
100g
Please to purchase this item. Need a web account? Register with us today!
10125403
View Documents Promotion Details
Thermo Scientific Acros
193275000
500 g Glass bottle
£392.96
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

10025-84-0
371.36
FDFPDGIMPRFRJP-UHFFFAOYSA-K
O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
Cl3H14LaO7
MFCD00149756
lanthanum(3+) heptahydrate trichloride

Specifications

91.0°C
White
99.99%
LaCl3·7H2O
15, 5414
FDFPDGIMPRFRJP-UHFFFAOYSA-K
lanthanum(3+) heptahydrate trichloride
371.38
Trace Metal Basis
10025-84-0
Crystalline Powder, Crystals and/or Chunks
Cl3H14LaO7
MFCD00149756
Solubility in water: soluble. Other solubilities: soluble in acids and alcohols
O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
371.36
99.99%
Lanthanum chloride heptahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only