missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Chloride Heptahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7073253 Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10457260

Brand: Fisher Chemical L/0451/46

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cl3La.7H2O, CAS Number-10025-84-0, 100g, White, 371.38g/mol, 91 deg.C, 149756, Plastic powder jar, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]

Specifications

Melting Point 91°C
Color White
Physical Form Solid
Quantity 25 g
Formula Weight 371.38g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lanthanum chloride heptahydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.