Learn More
Lanthanum chloride, hydrated, 64.5-70% LaCl3, Thermo Scientific Chemicals
£42.21 - £622.10
Chemical Identifiers
CAS | 20211-76-1 |
---|---|
Molecular Formula | Cl3La |
Molecular Weight (g/mol) | 245.26 |
MDL Number | MFCD00149753 |
InChI Key | ICAKDTKJOYSXGC-UHFFFAOYSA-K |
IUPAC Name | lanthanum(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[La+3] |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10144360
|
Thermo Scientific Acros
199151000 |
100 g | Glass bottle |
£42.21
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10114410
|
Thermo Scientific Acros
199155000 |
500 g | Glass bottle |
£149.75
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974191
|
Thermo Scientific Acros
199150025 |
2.5 kg | Glass bottle |
£622.10
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
20211-76-1 | |
245.26 | |
ICAKDTKJOYSXGC-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[La+3] |
Cl3La | |
MFCD00149753 | |
lanthanum(3+) trichloride |
Spezifikation
20211-76-1 | |
Adhering Crystals | |
LaCl3·xH2O | |
10,5193 | |
ICAKDTKJOYSXGC-UHFFFAOYSA-K | |
lanthanum(3+) trichloride | |
245.27 | |
Lanthanum chloride, hydrated |
White | |
Cl3La | |
MFCD00149753 | |
Solubility in water: soluble | |
[Cl-].[Cl-].[Cl-].[La+3] | |
245.26 | |
64.5 to 70% LaCl3 |
Sicherheit und Handhabung
GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
May be corrosive to metals.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
- Lanthanum chloride, hydrated
Signalwort
- Danger
Gefahrenkategorie
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
Gefahrenhinweis
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.