Learn More
Lanthanum chloride, hydrated, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 20211-76-1 |
Color | White |
Physical Form | Adhering Crystalline Powder or Crystals |
Quantity | 50 g |
Molecular Formula | Cl3La |
Linear Formula | LaCl3·xH2O |
MDL Number | MFCD00149753 |
Merck Index | 14, 5363 |
Solubility Information | Solubility in water: soluble |
InChI Key | ICAKDTKJOYSXGC-UHFFFAOYSA-K |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464727 | Certificate of Analysis | 09/08/2024 | 413010500 |
A0462676 | Certificate of Analysis | 15/05/2024 | 413010500 |
A0460974 | Certificate of Analysis | 14/03/2024 | 413010500 |
A0447360 | Certificate of Analysis | 09/11/2022 | 413010500 |
A0434685 | Certificate of Analysis | 11/10/2021 | 413010500 |
Safety and Handling



- Lanthanum chloride, hydrated
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.