missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) acetate sesquihydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11376328
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25721-92-0
Molecular Formula C12H24La2O15
Molecular Weight (g/mol) 686.12
MDL Number MFCD00013037
InChI Key RBPQRWPSWRTKEC-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) hexaacetate trihydrate
SMILES O.O.O.[La+3].[La+3].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point <200°C (in -1.5H2O)
Physical Form Crystalline
Quantity 50 g
Linear Formula La(OOCCH3)3·1.5H2O
Formula Weight 343.05 (316.03 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) acetate sesquihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'