missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11338697
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a scintillation material in certain application, as a source in the synthesis of lanthanum & its derivatives CsNaLa6Br14N2 and La3Br6N which contain rare bitetrahedral structural units..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-79-3
Molecular Formula Br3La
Molecular Weight (g/mol) 378.62
MDL Number MFCD00016161
InChI Key XKUYOJZZLGFZTC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) tribromide
SMILES [Br-].[Br-].[Br-].[La+3]

Specifications

Physical Form Beads
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 378.62
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) bromide, Ultra dry

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ27K053 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number035800.09,035800.04,035800.18
Lot NumberM06H029 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number035800.09,035800.04,035800.18
Lot NumberG11R012 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number035800.09,035800.04,035800.18
Lot NumberK18N33 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number035800.09,035800.04,035800.18
Lot NumberJ07M52 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number035800.09,035800.04,035800.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.