missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11347198

Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11347198
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11347198

Brand: Thermo Scientific Alfa Aesar 016639.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in pharmaceuticals & used in medicine as a phosphate binder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54451-24-0
Molecular Formula C3La2O9
Molecular Weight (g/mol) 457.84
MDL Number MFCD00011069
InChI Key NZPIUJUFIFZSPW-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) tricarbonate
SMILES [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Powder
Quantity 100 g
Merck Index 14,5364
Solubility Information Insoluble in water.
Formula Weight 457.85 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Density 2.6 g/mL to 2.7 g/mL
Assay (REO)
Chemical Name or Material Lanthanum(III) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.