missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals Product Code.: 11483047

Lanthanum(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals

Product Code. 11483047
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11483047

Brand: Thermo Scientific Alfa Aesar A15575.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

Solubility
Soluble in ethanol and water.

Notes
Incompatible with oxidizing agents, strong acids, strong reducing agents and active metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]

Specifications

Melting Point 91°C (decomposition)
Quantity 100 g
Merck Index 14,5363
Solubility Information Soluble in ethanol and water.
Formula Weight 371.37 (245.27 Anhydrous)
Percent Purity 99%
Odor Odorless
Chemical Name or Material Lanthanum(III) chloride heptahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250971 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15575.0E,A15575.22,A15575.36
Lot Number10245256 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog NumberA15575.0E,A15575.22,A15575.36
Lot Number10239572 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15575.0E,A15575.22,A15575.36
Lot Number10166327 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15575.0E,A15575.22,A15575.36
Lot Number10208142 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15575.0E,A15575.22,A15575.36
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lanthanum(III) chloride heptahydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Lanthanum(III) chloride heptahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.