missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(Iii) Chloride Heptahydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 25 g |
Linear Formula | LaCl3 · 7H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 371.37g/mol |
Grade | ACS Reagent |
Chemical Name or Material | Lanthanum(Iii) Chloride Heptahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'