missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) chloride heptahydrate, REacton, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 91°C (decomposition) |
Color | White |
Physical Form | Wet Crystalline Aggregates |
Quantity | 250 g |
Assay Percent Range | 99.99% (REO) |
Solubility Information | Soluble in water,alcohol and acids. |
Formula Weight | 371.37 (245.27 Anhydrous) |
Odor | Odorless |
Appearance | White wet crystalline aggregates |
Chemical Name or Material | Lanthanum(III) chloride heptahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'