missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride hydrate, 99.9% (REO), Thermo Scientific Chemicals

£52.76 - £194.97

Chemical Identifiers

CAS 20211-76-1
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00149753
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374788
View Documents
Thermo Scientific Alfa Aesar
043120.22
100 g
£52.76
100g
Please to purchase this item. Need a web account? Register with us today!
11384788
View Documents
Thermo Scientific Alfa Aesar
043120.36
500 g
£194.97
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lanthanum(III) chloride is useful as precursor for synthesis of aqueous sol-gel lanthanum phosphate nano rods,as a filter aid,an effective flocculent and precipitation phosphate. It is also used as a catalyst for the high pressure oxidative chlorination of methane to chloromethane and organic synthesis it acts as Lewis acid catalyst. It is used as a scintillator material and in Gamma Detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20211-76-1
245.26
ICAKDTKJOYSXGC-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[La+3]
Cl3La
MFCD00149753
lanthanum(3+) trichloride

Specifications

91°C (decomposition)
5
Cl3La
14,5363
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
245.27 (Anhydrous)
Odorless
Lanthanum (III) chloride hydrate
20211-76-1
Crystalline
MFCD00149753
Soluble in water and ethanol.
[Cl-].[Cl-].[Cl-].[La+3]
245.26
99.9%
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c

H290-H317-H318

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 233-237-5

missing translation for 'rtecsNumber' : OE4470000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only