missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

For vacuum deposition

£57.79 - £222.11

Chemical Identifiers

CAS 13709-38-1
Molecular Formula F3La
Molecular Weight (g/mol) 195.90
MDL Number MFCD00011073
InChI Key BYMUNNMMXKDFEZ-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trifluoride
SMILES [F-].[F-].[F-].[La+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354118
View Documents
Thermo Scientific Alfa Aesar
041414.09
10 g
£57.79
10g
Please to purchase this item. Need a web account? Register with us today!
11344118
View Documents
Thermo Scientific Alfa Aesar
041414.18
50 g
£222.11
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lanthanum fluoride, anhydrous is employed extensively in the preparation and study of fluoride glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13709-38-1
195.90
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
[F-].[F-].[F-].[La+3]
F3La
MFCD00011073
lanthanum(3+) trifluoride

Specifications

1493°C
Pieces
LaF3
Soluble in strong mineral acids. Insoluble in water.
[F-].[F-].[F-].[La+3]
195.90
99.9%
Hygroscopic
(REO)
13709-38-1
F3La
MFCD00011073
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
lanthanum(3+) trifluoride
195.9
Odorless
5.936 g/mL
Lanthanum(III) fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-252-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only