missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11316928
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Lanthanum fluoride, anhydrous is employed extensively in the preparation and study of fluoride glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-38-1
Molecular Formula F3La
Molecular Weight (g/mol) 195.90
MDL Number MFCD00011073
InChI Key BYMUNNMMXKDFEZ-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trifluoride
SMILES [F-].[F-].[F-].[La+3]

Specifications

Melting Point 1493°C
Physical Form Lump
Quantity 50 g
Linear Formula LaF3
Solubility Information Insoluble in water. Soluble in strong mineral acids.
Formula Weight 195.9
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 5.936 g/mL
Assay (REO)
Chemical Name or Material Lanthanum(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'