missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) hydroxide, 99.95% (REO), Thermo Scientific Chemicals Product Code.: 11314798

Lanthanum(III) hydroxide, 99.95% (REO), Thermo Scientific Chemicals

Product Code. 11314798
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11314798

Brand: Thermo Scientific Alfa Aesar 043123.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum(III) hydroxide is used in three way catalyst, glass, ceramic and electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14507-19-8
Molecular Formula La(OH)3
MDL Number MFCD00016160
Synonym lanthanum hydroxide, la oh 3, lanthanum hydroxide van, lanthanum 3+ trihydroxide, acmc-1c707, lanthanum 3+ ion trihydroxide, lanthanum iii hydroxide trace rare earth metals basis 50g

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water. Soluble in strong acids.
Formula Weight 189.93
Percent Purity 99.95%
Odor Odorless
Assay (REO)
Chemical Name or Material Lanthanum (III) hydroxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR16F039 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number043123.18,043123.30
Lot NumberS06D013 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number043123.18,043123.30,043123.A1
Lot NumberX05J051 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number043123.18,043123.30,043123.A1
Lot NumberY16H033 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043123.18,043123.30
Lot NumberC10P18 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number043123.18,043123.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) hydroxide, 99.95% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.