missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11358697
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13813-22-4
I3La
519.62
MFCD00049470
KYKBXWMMXCGRBA-UHFFFAOYSA-K
lanthanum(3+) triiodide
[I-].[I-].[I-].[La+3]
This item is not returnable. View return policy
13813-22-4
I3La
519.62
MFCD00049470
KYKBXWMMXCGRBA-UHFFFAOYSA-K
lanthanum(3+) triiodide
[I-].[I-].[I-].[La+3]

Lanthanum(III) iodide is used as a catalyst in the transesterification of N-acyloxazolidinones with alcohol gives the corresponding esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 772°C
Physical Form Beads
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 519.62
Percent Purity 99.9%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'