missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals Product Code.: 10571081

Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals

Product Code. 10571081
500 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10571081

Brand: Thermo Scientific Acros 218715000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronateShow MoreShow Less
PubChem CID 16211478
IUPAC Name lanthanum(3+) hexahydrate trinitrate
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 126.0°C
Melting Point 40.0°C
Color White to Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 98% min. (Complexometry)
Linear Formula La(NO3)3·6H2O
Fieser 11,283
Merck Index 15, 5414
Solubility Information Solubility in water: 1580g/L (25°C). Other solubilities: soluble in acids, soluble in alcohol
Formula Weight 433.01
Percent Purity ≥98%
Packaging Glass bottle
Chemical Name or Material Lanthanum(III) nitrate hexahydrate, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462578 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number218715000,218711000
Lot NumberA0459061 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number218715000,218711000
Lot NumberA0402616 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number218715000,218711000
Lot NumberA0419306 Certificate TypeCertificate of Analysis Date14/11/2022 Catalog Number218715000,218711000
Lot NumberA0438664 Certificate TypeCertificate of Analysis Date02/02/2022 Catalog Number218715000,218711000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.