missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10524084

Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10524084
500 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10524084

Brand: Thermo Scientific Acros 193285000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate
PubChem CID 16211478
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 126.0°C
Melting Point 40.0°C
Color Gray
Physical Form Powder/Granules
Quantity 500 g
Assay Percent Range 99.995%
Linear Formula La(NO3)3·6H2O
Fieser 11,283
Merck Index 15, 5414
Solubility Information Solubility in water: 1580g/L (25°C). Other solubilities: soluble in acids,soluble in alcohol
IUPAC Name lanthanum(3+);trinitrate;hexahydrate
Formula Weight 433.01
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467161 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number193280050,193285000,193281000
Lot NumberA0460475 Certificate TypeCertificate of Analysis Date26/02/2024 Catalog Number193280050,193285000,193281000
Lot NumberA0458024 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog Number193280050,193285000,193281000
Lot NumberA0453874 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number193280050,193285000,193281000
Lot NumberA0413126 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number193280050,193285000,193281000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.