Learn More
Lanthanum(III) nitride, 99.9% (REO), Thermo Scientific Chemicals
£108.00 - £423.00
Chemical Identifiers
CAS | 25764-10-7 |
---|---|
Molecular Formula | LaN |
Molecular Weight (g/mol) | 152.912 |
MDL Number | MFCD00049962 |
InChI Key | QCLQZCOGUCNIOC-UHFFFAOYSA-N |
Synonym | lanthanum nitride, lanthanum iii nitride, lanthanumylidyneamine |
PubChem CID | 117627 |
IUPAC Name | azanylidynelanthanum |
SMILES | N#[La] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358108
|
Thermo Scientific Alfa Aesar
039342.06 |
5 g |
£108.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348108
|
Thermo Scientific Alfa Aesar
039342.14 |
25 g |
£423.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lanthanum(III) nitride is used as a precursor along with silicon alkoxide to synthesize La9.33Si6O26, which finds application as electrolytes for solid oxide fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25764-10-7 | |
152.912 | |
QCLQZCOGUCNIOC-UHFFFAOYSA-N | |
117627 | |
N#[La] |
LaN | |
MFCD00049962 | |
lanthanum nitride, lanthanum iii nitride, lanthanumylidyneamine | |
azanylidynelanthanum |
Specifications
25764-10-7 | |
LaN | |
lanthanum nitride, lanthanum iii nitride, lanthanumylidyneamine | |
QCLQZCOGUCNIOC-UHFFFAOYSA-N | |
azanylidynelanthanum | |
117627 | |
99.9% | |
Lanthanum(III) nitride |
Powder | |
MFCD00049962 | |
Insoluble in water. Soluble in acid. | |
N#[La] | |
152.912 | |
152.91 | |
(REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 247-245-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only