missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxalate hydrate, 99.9% (REO), Thermo Scientific Chemicals

£54.57 - £202.01

Chemische Identifikatoren

CAS 79079-18-8
Molecular Formula C6La2O12
Molecular Weight (g/mol) 541.87
MDL Number MFCD00150540
InChI Key OXHNIMPTBAKYRS-UHFFFAOYSA-H
Synonym lanthanum iii oxalate decahydrate, lanthanum oxalate decahydrate, lanthanum iii oxalate, bis 4,5-dioxo-1,3-dioxa-2-lanthanacyclopentan-2-yl oxalate decahydrate
PubChem CID 91886560
IUPAC Name lanthanum;oxalic acid;decahydrate
SMILES [La+3].[La+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11304798
Unterlagen ansehen
Thermo Scientific Alfa Aesar
043122.22
100 g
£54.57
100g
Please to purchase this item. Need a web account? Register with us today!
11394788
Unterlagen ansehen
Thermo Scientific Alfa Aesar
043122.36
500 g
£202.01
500g
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

Lanthanum Oxalate is mainly applied in specialty glass, phosphors, water treatment and catalyst. This material use in glass, ceramics, paper, textile, coating, fine and specialty chemicals. In petroleum cracking catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

79079-18-8
541.87
OXHNIMPTBAKYRS-UHFFFAOYSA-H
91886560
[La+3].[La+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
C6La2O12
MFCD00150540
lanthanum iii oxalate decahydrate, lanthanum oxalate decahydrate, lanthanum iii oxalate, bis 4,5-dioxo-1,3-dioxa-2-lanthanacyclopentan-2-yl oxalate decahydrate
lanthanum;oxalic acid;decahydrate

Spezifikation

(decomposition)
Powder
La2(C2O4)3·xH2O
UN3288
Soluble in water. Soluble in strong mineral acids.
[La+3].[La+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
541.87
541.87 (Anhydrous)
Lanthanum (III) oxalate hydrate
79079-18-8
C6La2O12
MFCD00150540
lanthanum iii oxalate decahydrate, lanthanum oxalate decahydrate, lanthanum iii oxalate, bis 4,5-dioxo-1,3-dioxa-2-lanthanacyclopentan-2-yl oxalate decahydrate
OXHNIMPTBAKYRS-UHFFFAOYSA-H
lanthanum;oxalic acid;decahydrate
91886560
Hygroscopic
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 208-656-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

RUO – Research Use Only