missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Lanthanum(III) oxide is used for vacuum deposition. It is also acts as a precursor to LAMOX fast ion conductors. It is also employed in the manufacture of special alloys, optical glass and refractory materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 4200°C |
Melting Point | 2307°C |
Physical Form | Sintered Lump |
Quantity | 50 g |
Merck Index | 14,5363 |
Solubility Information | Insoluble in water; Soluble in dilute mineral acids |
Formula Weight | 325.82 |
Percent Purity | 99.9% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'