missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade, Thermo Scientific Chemicals Product Code.: 11346338

Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade, Thermo Scientific Chemicals

Product Code. 11346338
50 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11346338

Brand: Thermo Scientific Alfa Aesar 011266.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacturing of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
MDL Number MFCD00011071

Specifications

Boiling Point 4200°C
Melting Point 2307°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water,but soluble in dilute acid.
Formula Weight 325.82
Grade AAS
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder/lump
Density 6.51
Chemical Name or Material Lanthanum(III) oxide, For AAS analysis
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW07K015 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number011266.18,011266.A1,011266.30
Lot NumberU06K030 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number011266.18,011266.A1,011266.30
Lot NumberX19I030 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number011266.18,011266.A1,011266.30
Lot NumberW06J058 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number011266.18,011266.A1,011266.30
Lot NumberU15J031 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog Number011266.18,011266.A1,011266.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.