Learn More
Lanthanum(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
£70.50 - £400.00
Chemical Identifiers
CAS | 1312-81-8 |
---|---|
Molecular Formula | La2O3 |
Molecular Weight (g/mol) | 325.82 |
MDL Number | MFCD00011071 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10224751
|
Thermo Scientific Acros
193291000 |
100 g | Glass bottle |
£70.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137413
|
Thermo Scientific Acros
193295000 |
500 g | Glass bottle |
£236.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13418438
|
Thermo Scientific Acros
193290010 |
1 kg | Glass bottle |
£400.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
1312-81-8 | |
La2O3 | |
325.82 | |
MFCD00011071 |
Specifications
4200.0°C | |
1312-81-8 | |
Mass, Crystals, Crystalline Powder and/or Chunks | |
La2O3 | |
MFCD00011071 | |
Solubility in water: insoluble | |
325.82 | |
Trace Metal Basis | |
Lanthanum(III) oxide |
2315.0°C | |
White to Yellow | |
99.94% min. (La2O3) TREO base | |
La2O3 | |
15,5414 | |
325.82 | |
99.99% | |
2.0 % max. (1000°C) |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 215-200-5
RUO – Research Use Only