missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, REacton™, 99.9% (REO)

Product Code. p-4407514
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Produktkod. 11366348

Brand: Thermo Scientific Alfa Aesar 011272.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 1312-81-8 | La2O3

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacture of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1312-81-8
MDL Number MFCD00011071
Synonym Lanthanum trioxide, Lanthanum sesquioxide

Specifikationer

Boiling Point 4200°C
Melting Point 2307°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (REO)
Molecular Formula La2O3
Solubility Information Insoluble in water but soluble in dilute acid.
Formula Weight 325.82
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder/lump
Density 6.51 g/cm3 at 20°C
Chemical Name or Material Lanthanum(III) oxide
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.