missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) perchlorate hexahydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Lanthanum(III) perchlorate hexahydrate participates in the preparation of the Schiff base complexes of lanthanide (III) ions. When hydrated lanthanum perchlorate in ethanolic solution is treated with an ethanolic solution of terpyridine, [La(terpy)3](CIO4)3 is deposited as very small crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Crystalline |
Quantity | 25 g |
UN Number | UN1481 |
Solubility Information | Easily soluble in water. |
Formula Weight | 545.35 (437.26 Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Chemical Name or Material | Lanthanum(III) perchlorate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'