missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) sulfate hydrate, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11374768

Lanthanum(III) sulfate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11374768
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11374768

Brand: Thermo Scientific Alfa Aesar 043106.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as colorant or laboratory reagent. It is also mainly applied in specialty glass, phosphors, water treatment and catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57804-25-8
Molecular Formula La2O12S3
Molecular Weight (g/mol) 565.98
MDL Number MFCD00011070
InChI Key VQEHIYWBGOJJDM-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) trisulfate
SMILES [La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point White heat (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (REO)
Solubility Information Slightly soluble in water.
Formula Weight 566.00 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.821
Chemical Name or Material Lanthanum(III) sulfate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR30J029 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog Number043106.36,043106,043106.22
Lot NumberK28S036 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043106.36,043106.22
Lot NumberK18T050 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043106.36,043106.22
Lot NumberP13F048 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043106.36,043106.22
Lot NumberY18F047 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043106.36,043106.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) sulfate hydrate, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.