Learn More
Lanthanum nickel powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
£64.52 - £258.29
Chemical Identifiers
CAS | 12196-72-4 |
---|---|
Molecular Formula | LaNi5 |
Molecular Weight (g/mol) | 432.37 |
MDL Number | MFCD00198177 |
InChI Key | BUCVQDCWULHLJZ-UHFFFAOYSA-N |
Synonym | lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloyShow More |
PubChem CID | 14367487 |
IUPAC Name | lanthanum;nickel |
SMILES | [Ni].[Ni].[Ni].[Ni].[Ni].[La] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369389
|
Thermo Scientific Alfa Aesar
089133.06 |
5 g |
£64.52
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359389
|
Thermo Scientific Alfa Aesar
089133.14 |
25 g |
£258.29
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
12196-72-4 | |
432.37 | |
BUCVQDCWULHLJZ-UHFFFAOYSA-N | |
14367487 | |
[Ni].[Ni].[Ni].[Ni].[Ni].[La] |
LaNi5 | |
MFCD00198177 | |
lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloyShow More | |
lanthanum;nickel |
Specifications
12196-72-4 | |
MFCD00198177 | |
lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy | |
[Ni].[Ni].[Ni].[Ni].[Ni].[La] | |
432.37 | |
99.5% | |
Odorless | |
(metals basis) |
LaNi5 | |
UN3178 | |
BUCVQDCWULHLJZ-UHFFFAOYSA-N | |
lanthanum;nickel | |
14367487 | |
−100 Mesh | |
Air and moisture sensitive | |
Lanthanum nickel powder |
Safety and Handling
GHS H Statement
H228-H260-H350-H317
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P210-P240-P241-P260-P264b-P270-P281-P285-P304+P341-P308+P313-P342+P311-P370+P378q-P501c
H228-H334-H350-H372
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III
missing translation for 'einecsNumber' : 235-372-5
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.