missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum nickel, REacton™, 99.9% (REO)

Product Code. p-7048697
Click to view available options
Quantity:
50 g
5 x 50 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11356348

Brand: Thermo Scientific Alfa Aesar 011271.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12196-72-4 | LaNi5 | 432.37 g/mol

Lanthanum nickel act as a superconducting intermetallic phase in lanthanum nickel boro-nitride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12196-72-4
Molecular Formula LaNi5
Molecular Weight (g/mol) 432.37
MDL Number MFCD00198177
InChI Key BUCVQDCWULHLJZ-UHFFFAOYSA-N
Synonym lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy
PubChem CID 14367487
IUPAC Name lanthanum;nickel
SMILES [Ni].[Ni].[Ni].[Ni].[Ni].[La]

Specifications

Quantity 50 g
Odor Odorless
Sensitivity Air and moisture sensitive
Form Pieces
UN Number UN3178
Solubility Information Reacts with water at 0°C.
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Lanthanum nickel
compliance-icons compliance-icons
Product Identifier
  • Lanthanum, compound with nickel (1:5)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H228-Flammable solid.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Lanthanum, compoundwith nickel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.