missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum nitrate solution, Honeywell™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations

Specifications
Specifications
CAS | 10099-59-9 |
Quantity | 1 L |
Linear Formula | La(NO3)3 |
MDL Number | MFCD00011067 |
UN Number | NONH for all modes of transport |
Formula Weight | 324.92g/mol |
Chemical Name or Material | Lanthanum nitrate solution |
Safety and Handling

Product Identifier
- Lanthanum nitrate solution
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Lanthanum trinitrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lanthanum nitrate solution, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction