missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum Oxide, Extra Pure, SLR, Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Specifications
Boiling Point | 4200°C |
Melting Point | 2315°C |
CAS | 1312-81-8 |
Color | White |
Physical Form | Powder/Solid |
Quantity | 100 g |
Molecular Formula | La2O3 |
MDL Number | 11071 |
Formula Weight | 325.82g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'