Learn More
Lanthanum pieces, 99.9% (REO), Thermo Scientific Chemicals
£128.64 - £347.73
Chemical Identifiers
CAS | 7439-91-0 |
---|---|
Molecular Formula | La |
Molecular Weight (g/mol) | 138.91 |
MDL Number | MFCD00011066 |
InChI Key | FZLIPJUXYLNCLC-UHFFFAOYSA-N |
Synonym | in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation |
PubChem CID | 23926 |
ChEBI | CHEBI:33336 |
IUPAC Name | lanthanum |
SMILES | [La] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11390689
|
Thermo Scientific Alfa Aesar
000175.14 |
25 g |
£128.64
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11380689
|
Thermo Scientific Alfa Aesar
000175.22 |
100 g |
£347.73
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-91-0 | |
138.91 | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
23926 | |
lanthanum |
La | |
MFCD00011066 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
CHEBI:33336 | |
[La] |
Specifications
Silver-White | |
Silvery-white pieces | |
99.9% (REO) | |
MFCD00011066 | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
lanthanum | |
23926 | |
Under Mineral Oil |
7439-91-0 | |
6.17g/cm3 at 20°C | |
La | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
[La] | |
138.91 | |
CHEBI:33336 | |
Lanthanum, Rare Earth Oxides Content |
Safety and Handling
Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: II
missing translation for 'einecsNumber' : 231-099-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lanthanum pieces
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H228-Flammable solid.
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
- P501-Dispose of contents/container to .
RUO – Research Use Only