missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11341449
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8, 7697-37-2, 7732-18-5
Molecular Formula La2O3 in 5% HNO3
MDL Number MFCD00011066

Specifications

Quantity 50 mL
Color Colorless
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Lanthanum(III) oxide: 0.12%; Water: 94.88%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Lanthanum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'