missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO), Thermo Scientific Chemicals

Form: Approx.0.20g/mm, Cast rod, diameter is approximate

£84.22 - £424.11

Chemical Identifiers

CAS 7439-91-0
Molecular Formula La
Molecular Weight (g/mol) 138.91
MDL Number MFCD00011066
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
Synonym in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
PubChem CID 23926
ChEBI CHEBI:33336
IUPAC Name lanthanum
SMILES [La]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313828
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040292.HF
10 mm
£84.22
10mm
Please to purchase this item. Need a web account? Register with us today!
11343828
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040292.DH
25 mm
£150.75
25mm
Please to purchase this item. Need a web account? Register with us today!
11333828
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040292.DJ
50 mm
£240.20
50mm
Please to purchase this item. Need a web account? Register with us today!
11323828
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040292.DM
100 mm
£424.11
100mm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-91-0
138.91
FZLIPJUXYLNCLC-UHFFFAOYSA-N
23926
lanthanum
La
MFCD00011066
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
CHEBI:33336
[La]

Specifications

Silver-White
Silvery-white rod
La
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
[La]
138.91
CHEBI:33336
Lanthanum, Rare Earth Oxides Content
7439-91-0
99.9% (REO)
MFCD00011066
FZLIPJUXYLNCLC-UHFFFAOYSA-N
lanthanum
23926
Cast Rod
Videos
Safety and Handling

Safety and Handling

Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: II

missing translation for 'einecsNumber' : 231-099-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only