missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum sputtering target, 50.8mm dia x 3.18mm thick, Thermo Scientific™ Product Code.: 15407065

Lanthanum sputtering target, 50.8mm dia x 3.18mm thick, Thermo Scientific™

Product Code. 15407065
1 Ea.
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy

Product Code. 15407065

Brand: Thermo Scientific Alfa Aesar 045740.KS

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
CAS 7439-91-0
Molecular Formula La
MDL Number MFCD00011066
UN Number UN3208
Quantity 1 Ea.
Synonym in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
SMILES [La]
IUPAC Name lanthanum
Molecular Weight (g/mol) 138.91
PubChem CID 23926
ChEBI CHEBI:33336
Chemical Name or Material Lanthanum sputtering target, 50.8mm dia. x 3.18mm thick
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum sputtering target, 50.8mm dia x 3.18mm thick, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.