missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Cellular Assay Lysis Buffer (6X)
LanthaScreen™ Cellular Assay Lysis Buffer (6X)
Brand: Invitrogen A12891
Code : 50
Additional Details : Weight : 0.99000kg
Description
LanthaScreen™ 6X Cellular Assay Lysis buffer is specially formulated for use with LanthaScreen™ Cellular Assays.Specifications
Cellular Assay Lysis Buffer | |
6X | |
LanthaScreen Cellular Assays | |
6 mL |
LanthaScreen™ Cellular Assay Lysis Buffer, 6mL, Liquid, 6X Concentration, 4°C Storage, For use with Pharma and Biopharma, Target and Lead Identification and Validation, LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays | |
Pharma and biopharma, target and lead identification and validation | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.