missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Cellular Assay Lysis Buffer
LanthaScreen™ Cellular Assay Lysis Buffer
Brand: Invitrogen PV5598
Code : 50
Additional Details : Weight : 0.10000kg
Description
LanthaScreen™ Cellular Assay Lysis buffer is specially formulated for use with the LanthaScreen™ Cellular Assays.Specifications
Cell Lysis Buffer | |
20 mM (Tris HCl), 150 mM (NaCl), 500 mM (NaF), 5 mM (EDTA) and 1% (NP-40) | |
Liquid | |
20mM Tris HCl (pH 7.5), 150mM NaCl, 500mM NaF, 5mM EDTA, and 1% NP-40 |
LanthaScreen™ Cellular Assay Lysis Buffer, 100mL, Liquid, Store at 4°C, Cell Lysis Buffer, Used For Pharma & Biopharma, Target & Lead Identification & Validation | |
Pharma and biopharma, target and lead identification and validation, drug discovery and development, kinase biology, kinase pathway analysis, pathway biology, cellular pathway analysis assays, other cellular pathway assay products | |
100 mL |
For Research Use Only. Not for use in diagnostic procedures.