Learn More
Thermo Scientific™ LanthaScreen™ Eu-PY20 Antibody Kit
LanthaScreen™ Eu-PY20 Antibody Kit
£762.00 - £8520.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10013954
|
Thermo Scientific™
PV5692 |
25μg |
£762.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10003624
|
Thermo Scientific™
PV5696 |
1 mg |
£8520.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A general reagent to monitor tyrosine kinase activity, the PY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate. For use in combination with AlexaFluor 647-poly-GT (PV5693) to evaluate inhibitors in medium to high throughput screening applications.For Research Use Only. Not for use in diagnostic procedures.