missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Tb-Anti-Mouse Antibody
LanthaScreen™ Tb-Anti-Mouse Antibody
Brand: Invitrogen™ PV5264
Code : 50
Additional Details : Weight : 0.50000kg
Description
LanthaScreen™ Tb-labeled phospho-specific antibodies, allow for the detection of intracellular post-translational modifications of specific target proteins. To facilitate in-house kinase assay configuration and screening, we have developed a selection of Tb-labeled phospho-specific antibodies, which may be accessed in small scale and screening quantities.Specifications
Lanthanide Chelates, Terbium (Tb) | |
-5°C to -30°C | |
50 μg | |
Liquid | |
LanthaScreen™ Tb-Anti-Mouse Antibody |
LanthaScreen™ Tb-labeled Anti-Mouse Secondary Antibody, 50μg, Liquid, 1.03mg/mL Concentration, Mouse Species Reactivity, Rabbit Host, IgG Isotype, Terbium Conjugate, Fluorescent Detection Method, 6 Months Shelf Life, Store in Freezer, Used for Ph | |
Fluorescent | |
Liquid | |
Pharma and biopharma, secondary detection, cell analysis, drug discovery and development, target and lead identification and validation, kinase biology, kinase antibodies and immunoassays, biochemical kinase assays, secondary detection |
For Research Use Only. Not for use in diagnostic procedures.