missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-labeled Anti-Goat Antibody Kit

Product Code. p-7181444
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
1mg
25µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10779544

Brand: Thermo Scientific™ PV3771

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents to facilitate in-house assay development around multiple target classes and assay formats.

  • Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents to facilitate in-house assay development around multiple target classes and assay formats
  • Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species
  • Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control
  • In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled
  • The detection step is performed by the addition of the Tb-labeled secondary antibody
  • This approach can be applied to other target classes and assay formats
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Quantity 1 mg
Reactivity Goat
Shipping Condition Dry Ice
Conjugate Lanthanide Chelates, Terbium (Tb)
Product Line LanthaScreen

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.