missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-labeled Anti-Mouse Ab

Product Code. p-7182869
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10309204

Brand: Invitrogen™ PV3766

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

LanthaScreen™ Tb-labeled Anti-Mouse Ab

A general reagent for use in assay development, this terbium labeled antibody will bind mouse antibodies. This product is ideally suited for use during assay development when evaluating multiple primary mouse antibodies or when you are unable to directly terbium label your primary antibody.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Terbium-labeled anti-Mouse antibody in HEPES buffered saline. Store at -20°C.
Excitation/Emission 340/490, 520
Quantity 25 μg
Reactivity Mouse
Readout End Point
Shipping Condition Dry Ice
Conjugate Lanthanide Chelates, Terbium (Tb)
Format Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.